siRNA Details
Gene Symbol: RRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102669.2 | NP_001096139.1 | 3, 4 | 466 | ||
NM_001177314.1 | NP_001170785.1 | 3, 4 | 311 | |||
NM_001177315.1 | NP_001170786.1 | 3, 4 | 443 | |||
NM_012250.5 | NP_036382.2 | 3, 4 | 617 | |||
XM_017017363.1 | XP_016872852.1 | 3, 4 | 282 | |||
GenBank mRNA | AF493924.1 | - | 3, 4 | 300 | ||
AK299606.1 | - | 4, 5 | 467 | |||
AK300103.1 | - | 3, 4 | 311 | |||
AK302033.1 | - | 3, 4 | 283 | |||
AK309550.1 | - | 3, 4 | 443 | |||
AK309683.1 | - | 3, 4 | 424 | |||
AK313976.1 | - | 3, 4 | 616 | |||
BC013106.1 | - | 3, 4 | 500 | 4389214 | ||
BG428983.1 | - | 3, 4 | 465 | |||
BX324335.2 | - | 3, 4 | 603 | |||
CB961515.1 | - | 604 | ||||
DC368033.1 | - | 3, 4 | 424 | |||
M31468.1 | - | 3, 4 | 300 |
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG028932.1 | - | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
