siRNA Details
Gene Symbol: NPHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153240.4 | NP_694972.3 | 1 | 396 | ||
GenBank mRNA | AK055893.1 | - | 1 | 369 | ||
AL832877.1 | - | 1 | 346 | |||
AY257864.1 | - | 1 | 325 | |||
BC068082.1 | - | 1 | 351 | |||
DB114822.1 | - | 1 | 379 |
Gene Symbol: NPHP3-ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037804.1 | 1 | 396 | |||
GenBank mRNA | AB082531.1 | - | 1 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
