siRNA Details
Gene Symbol: LINC00526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026849.1 | 1 | 681 | |||
GenBank mRNA | AK292743.1 | - | 1 | 1258 | ||
BC010538.1 | - | 1 | 681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
