siRNA Details
Gene Symbol: ZNF417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297734.1 | NP_001284663.1 | 3 | 2194 | ||
NM_152475.2 | NP_689688.2 | 3 | 2191 | |||
XM_011526473.2 | XP_011524775.1 | 3 | 2784 | |||
XM_017026315.1 | XP_016881804.1 | 3 | 2057 | |||
GenBank mRNA | AK293789.1 | - | 4 | 2350 | ||
BC025783.1 | - | 3 | 2157 | 5226312 |
