siRNA Details
Gene Symbol: IGF1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000875.4 | NP_000866.1 | 21 | 11949 | ||
NM_001291858.1 | NP_001278787.1 | 21 | 11946 | |||
XM_017022136.1 | XP_016877625.1 | 21 | 11452 | |||
XM_017022137.1 | XP_016877626.1 | 22 | 10966 | |||
XM_017022138.1 | XP_016877627.1 | 21 | 11449 | |||
GenBank mRNA | AB425196.1 | - | 21 | 11430 | ||
BC010607.1 | - | 1 | 2961 | |||
BC088377.1 | - | 1 | 980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
