siRNA Details
Gene Symbol: AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000044.3 | NP_000035.2 | 1 | 1459 | ||
GenBank mRNA | AF162704.1 | - | 2 | 326 | ||
AF321914.1 | - | 1 | 434 | |||
AF321915.1 | - | 1 | 422 | |||
AF321916.1 | - | 1 | 431 | |||
AF321917.1 | - | 2 | 398 | |||
BC132975.1 | - | 1 | 801 | 40146997 | ||
FJ235916.1 | - | 1 | 686 | |||
FJ235917.1 | - | 1 | 683 | |||
FJ235918.1 | - | 1 | 683 | |||
FJ235919.1 | - | 1 | 686 | |||
FJ235920.1 | - | 1 | 686 | |||
GU208210.1 | - | 1 | 350 | |||
HM055487.1 | - | 1 | 686 | |||
KJ534771.1 | - | 1 | 341 | |||
M20132.1 | - | 1 | 700 | |||
M21748.1 | - | 1 | 497 | |||
M23263.1 | - | 1 | 857 | |||
M34233.1 | - | 2 | 431 | |||
M73069.1 | - | 1 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
