siRNA Details
Gene Symbol: ZNF578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099694.1 | NP_001093164.1 | 6 | 1974 | ||
XM_017026302.1 | XP_016881791.1 | 6 | 1994 | |||
GenBank mRNA | AK055946.1 | - | 1 | 1152 | ||
AK095562.1 | - | 1 | 669 | |||
AK299106.1 | - | 6 | 1974 | |||
BC103904.2 | - | 1 | 1142 | |||
BC115704.1 | - | 1 | 1142 |
