siRNA Details
Gene Symbol: MEI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093341.1 | - | 7 | 1140 | ||
AK307341.1 | - | 3 | 828 | |||
AK311552.1 | - | 2 | 760 | |||
BC070111.1 | - | 23 | 3285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
