siRNA Details
Gene Symbol: VDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000376.2 | NP_000367.1 | 3, 4 | 304 | ||
NM_001017535.1 | NP_001017535.1 | 4, 5 | 426 | |||
NM_001017536.1 | NP_001017536.1 | 3, 4 | 695 | |||
XM_006719587.3 | XP_006719650.1 | 2, 3 | 221 | |||
XM_011538720.2 | XP_011537022.1 | 3, 4 | 336 | |||
GenBank mRNA | AB307700.1 | - | 1, 2 | 144 | ||
AF026260.1 | - | 1, 2 | 144 | |||
AK225844.1 | - | 3, 4 | 303 | |||
AK312267.1 | - | 3, 4 | 296 | |||
BC033465.1 | - | 3, 4 | 283 | |||
BC060832.1 | - | 3, 4 | 667 | 30343975 | ||
CF138099.1 | - | 3, 4 | 294 | |||
DA721726.1 | - | 4, 5 | 435 | |||
HQ692836.1 | - | 1, 2 | 144 | |||
J03258.1 | - | 3, 4 | 259 | |||
M65208.1 | - | 1, 2 | 75 | |||
X67482.1 | - | 2, 3 | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
