siRNA Details
Gene Symbol: TDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001578.1 | 8 | 1169 | |||
GenBank mRNA | AJ301562.1 | - | 8 | 1169 | ||
AK057762.1 | - | 8 | 1108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
