siRNA Details
Gene Symbol: HNF4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000457.4 | NP_000448.3 | 1 | 221 | ||
NM_001258355.1 | NP_001245284.1 | 1 | 221 | |||
NM_178849.2 | NP_849180.1 | 1 | 221 | |||
NM_178850.2 | NP_849181.1 | 1 | 221 | |||
XM_005260407.3 | XP_005260464.1 | 1 | 224 | |||
GenBank mRNA | AB307703.1 | - | 1 | 77 | ||
BC137539.1 | - | 1 | 143 | 9021539 | ||
BC137540.1 | - | 1 | 143 | 9021540 | ||
BP274413.1 | - | 1 | 221 | |||
HQ692860.1 | - | 1 | 104 | |||
HQ692869.1 | - | 1 | 104 | |||
X76930.1 | - | 1 | 96 | |||
X87870.1 | - | 1 | 176 | |||
X87871.1 | - | 1 | 161 | |||
X87872.1 | - | 1 | 197 | |||
Z49825.1 | - | 1 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
