siRNA Details
Gene Symbol: NR0B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000475.4 | NP_000466.2 | 1 | 1151 | ||
XM_017029338.1 | XP_016884827.1 | 1 | 1172 | |||
GenBank mRNA | BC011564.1 | - | 1 | 1148 | 4637694 | |
HQ692832.1 | - | 1 | 1136 | |||
S74720.1 | - | 1 | 1370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
