siRNA Details
Gene Symbol: FAM47C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013736.2 | NP_001013758.1 | 1 | 338 | ||
GenBank mRNA | AK125992.1 | - | 1 | 60 |
Gene Symbol: FAM47B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152631.2 | NP_689844.2 | 1 | 322 | ||
GenBank mRNA | AK093101.1 | - | 1 | 304 | ||
BC035026.2 | - | 1 | 322 | 4828379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
