siRNA Details
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025145.5 | NP_079421.5 | 29, 30 | 3876 | ||
XM_005270171.2 | XP_005270228.1 | 29, 30 | 3880 | |||
XM_011540196.2 | XP_011538498.1 | 30, 31 | 3994 | |||
XM_011540198.2 | XP_011538500.1 | 29, 30 | 3877 | |||
XM_011540199.2 | XP_011538501.1 | 29, 30 | 3877 | |||
XM_011540201.2 | XP_011538503.1 | 30, 31 | 3994 | |||
XM_011540202.2 | XP_011538504.1 | 26, 27 | 3310 | |||
XM_011540203.2 | XP_011538505.1 | 28, 29 | 3552 | |||
XM_017016681.1 | XP_016872170.1 | 30, 31 | 3991 | |||
XM_017016682.1 | XP_016872171.1 | 27, 28 | 3646 | |||
XM_017016683.1 | XP_016872172.1 | 27, 28 | 3432 | |||
GenBank mRNA | AK093325.1 | - | 3, 4 | 306 | ||
DA681478.1 | - | 4, 5 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
