siRNA Details
Gene Symbol: CCDC112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040440.2 | NP_001035530.1 | 5, 6 | 747 | ||
NM_152549.2 | NP_689762.2 | 5, 6 | 793 | |||
XM_005271908.1 | XP_005271965.1 | 5, 6 | 741 | |||
GenBank mRNA | AJ580807.1 | - | 5, 6 | 646 | ||
BC031242.1 | - | 5, 6 | 771 | 5265792 |
