siRNA Details
Gene Symbol: LEXM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110533.1 | NP_001104003.1 | 3 | 392 | ||
NM_152607.2 | NP_689820.2 | 3 | 392 | |||
XM_017000480.1 | XP_016855969.1 | 3 | 392 | |||
XM_017000481.1 | XP_016855970.1 | 3 | 392 | |||
GenBank mRNA | AK097520.1 | - | 3 | 392 | ||
BC039109.1 | - | 3 | 353 | 4837126 | ||
BC064142.1 | - | 3 | 354 | 30390069 |
