siRNA Details
Gene Symbol: DCLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040260.3 | NP_001035350.2 | 5 | 1798 | ||
NM_001040261.4 | NP_001035351.4 | 6 | 1849 | |||
NR_036614.1 | 5 | 1798 | ||||
XM_005262785.2 | XP_005262842.1 | 6 | 1678 | |||
XM_005262786.2 | XP_005262843.1 | 5 | 1627 | |||
XM_017007832.1 | XP_016863321.1 | 6 | 1678 | |||
XM_017007833.1 | XP_016863322.1 | 6 | 1678 | |||
GenBank mRNA | AB209181.1 | - | 5 | 1798 | ||
AL834498.1 | - | 6 | 896 | |||
BC032726.1 | - | 5 | 1091 | |||
DA201927.1 | - | 3 | 248 |
