siRNA Details
Gene Symbol: HMGCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000859.2 | NP_000850.1 | 2, 3 | 329 | ||
NM_001130996.1 | NP_001124468.1 | 2, 3 | 329 | |||
XM_011543357.1 | XP_011541659.1 | 2, 3 | 480 | |||
XM_011543358.1 | XP_011541660.1 | 2, 3 | 367 | |||
XM_011543359.1 | XP_011541661.1 | 2, 3 | 480 | |||
GenBank mRNA | AK292892.1 | - | 2, 3 | 260 | ||
AK296499.1 | - | 2, 3 | 485 | |||
AK312437.1 | - | 1, 2 | 173 | |||
BC033692.1 | - | 2, 3 | 265 | 5212903 | ||
M11058.1 | - | 2, 3 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
