siRNA Details
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025145.5 | NP_079421.5 | 30 | 3946 | ||
XM_005270171.2 | XP_005270228.1 | 30 | 3950 | |||
XM_011540196.2 | XP_011538498.1 | 31 | 4064 | |||
XM_011540198.2 | XP_011538500.1 | 30 | 3947 | |||
XM_011540199.2 | XP_011538501.1 | 30 | 3947 | |||
XM_011540201.2 | XP_011538503.1 | 31 | 4064 | |||
XM_011540202.2 | XP_011538504.1 | 27 | 3380 | |||
XM_011540203.2 | XP_011538505.1 | 29 | 3622 | |||
XM_017016681.1 | XP_016872170.1 | 31 | 4061 | |||
XM_017016682.1 | XP_016872171.1 | 28 | 3716 | |||
XM_017016683.1 | XP_016872172.1 | 28 | 3502 | |||
GenBank mRNA | AK093325.1 | - | 4 | 376 | ||
DA681478.1 | - | 5 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
