siRNA Details
Gene Symbol: CT45A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291527.1 | NP_001278456.1 | 5 | 900 | ||
NM_001291528.1 | NP_001278457.1 | 5 | 883 | |||
NM_001291529.1 | NP_001278458.1 | 5 | 821 | |||
NM_001291530.1 | NP_001278459.1 | 5 | 838 | |||
XM_011531238.2 | XP_011529540.1 | 5 | 834 | |||
XM_011531239.1 | XP_011529541.1 | 5 | 815 | |||
GenBank mRNA | AK292207.1 | - | 5 | 817 | ||
AY743713.1 | - | 5 | 976 | |||
BC070138.1 | - | 5 | 821 | 4685945 |
Gene Symbol: CT45A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017417.2 | NP_001017417.1 | 5 | 975 | ||
XM_005278141.3 | XP_005278198.1 | 5 | 824 | |||
XM_011531352.1 | XP_011529654.1 | 4 | 745 | |||
GenBank mRNA | AY743709.1 | - | 5 | 976 |
Gene Symbol: CT45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152582.6 | NP_689795.4 | 5 | 976 | ||
XM_006724783.2 | XP_006724846.1 | 5 | 903 | |||
XM_011531389.2 | XP_011529691.1 | 5 | 871 | |||
GenBank mRNA | AY743710.1 | - | 5 | 976 |
Gene Symbol: CT45A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017435.1 | NP_001017435.1 | 5 | 976 | ||
XM_006724758.3 | XP_006724821.1 | 5 | 893 | |||
XM_011531340.2 | XP_011529642.1 | 5 | 852 | |||
GenBank mRNA | AK098689.1 | - | 5 | 821 | ||
AY743711.1 | - | 5 | 976 |
Gene Symbol: CT45A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007551.4 | NP_001007552.2 | 5 | 976 | ||
NM_001172288.2 | NP_001165759.2 | 5 | 993 | |||
XM_011531341.2 | XP_011529643.1 | 5 | 858 | |||
GenBank mRNA | BC028711.2 | - | 5 | 976 | 4828274 |
Gene Symbol: CT45A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017438.2 | NP_001017438.2 | 5 | 976 | ||
XM_005262425.4 | XP_005262482.2 | 5 | 896 | |||
XM_011531351.1 | XP_011529653.1 | 5 | 827 | |||
GenBank mRNA | AY743714.1 | - | 5 | 976 |
Gene Symbol: CT45A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291543.1 | NP_001278472.1 | 5 | 792 | ||
XM_003960082.4 | XP_003960131.1 | 5 | 879 | |||
XM_011531229.2 | XP_011529531.1 | 5 | 858 | |||
GenBank mRNA | BU540102.1 | - | 5 | 796 |
Gene Symbol: CT45A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291535.1 | NP_001278464.1 | 5 | 976 | ||
XM_006724799.2 | XP_006724862.1 | 5 | 903 | |||
XM_011531241.2 | XP_011529543.1 | 5 | 871 |
Gene Symbol: CT45A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291540.2 | NP_001278469.1 | 5 | 976 | ||
NM_001321271.1 | NP_001308200.1 | 5 | 821 | |||
XM_006724796.2 | XP_006724859.1 | 5 | 903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
