siRNA Details
Gene Symbol: ZNF383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152604.1 | NP_689817.1 | 5 | 1553 | ||
XM_005258585.2 | XP_005258642.1 | 6 | 1811 | |||
XM_005258587.2 | XP_005258644.1 | 5 | 1688 | |||
XM_005258588.2 | XP_005258645.1 | 6 | 1450 | |||
XM_011526586.2 | XP_011524888.1 | 7 | 2556 | |||
XM_011526587.2 | XP_011524889.1 | 6 | 2523 | |||
XM_011526588.2 | XP_011524890.1 | 5 | 2705 | |||
XM_011526589.2 | XP_011524891.1 | 5 | 1386 | |||
XM_011526590.2 | XP_011524892.1 | 4 | 1351 | |||
XM_017026422.1 | XP_016881911.1 | 7 | 1962 | |||
XM_017026423.1 | XP_016881912.1 | 7 | 2556 | |||
XM_017026424.1 | XP_016881913.1 | 5 | 1602 | |||
GenBank mRNA | AK091760.1 | - | 5 | 2114 | ||
AK093182.1 | - | 5 | 1553 | |||
AK122892.1 | - | 8 | 1879 | |||
AY260141.1 | - | 4 | 1348 | |||
AY438646.1 | - | 5 | 1553 |

More Information
Other Species Targeted

Gene Ontology Categories:
