siRNA Details
Gene Symbol: KNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152643.6 | NP_689856.6 | 30 | 6368 | ||
GenBank mRNA | AK057756.1 | - | 4 | 1665 | ||
AK131205.1 | - | 11 | 2649 | |||
BC047924.1 | - | 11 | 2622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
