siRNA Details
Gene Symbol: RALGPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286247.1 | NP_001273176.1 | 13 | 1514 | ||
NM_152663.4 | NP_689876.2 | 13 | 1514 | |||
XM_006711410.3 | XP_006711473.1 | 13 | 1427 | |||
XM_006711411.3 | XP_006711474.1 | 6 | 644 | |||
XM_011509688.1 | XP_011507990.1 | 13 | 1428 | |||
XM_017001591.1 | XP_016857080.1 | 12 | 1261 | |||
GenBank mRNA | AK098470.1 | - | 2 | 192 | ||
AK301753.1 | - | 13 | 1420 | |||
BC047391.1 | - | 13 | 1499 | 5298449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
