siRNA Details
Gene Symbol: HIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152696.3 | NP_689909.2 | 2 | 272 | ||
NM_198268.2 | NP_938009.1 | 2 | 272 | |||
XM_005270609.3 | XP_005270666.1 | 2 | 449 | |||
XM_005270610.4 | XP_005270667.1 | 2 | 449 | |||
XM_005270611.4 | XP_005270668.1 | 2 | 449 | |||
XM_005270612.4 | XP_005270669.1 | 2 | 411 | |||
XM_005270613.4 | XP_005270670.1 | 4 | 362 | |||
XM_006710443.3 | XP_006710506.1 | 2 | 449 | |||
XM_011540977.2 | XP_011539279.1 | 2 | 449 | |||
XM_017000605.1 | XP_016856094.1 | 2 | 411 | |||
XM_017000606.1 | XP_016856095.1 | 2 | 449 | |||
GenBank mRNA | AB089957.1 | - | 2 | 236 | ||
AK122662.1 | - | 2 | 270 | |||
AK302726.1 | - | 2 | 456 | |||
BC033012.2 | - | 2 | 272 | 4830408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
