siRNA Details
Gene Symbol: DKFZp434L192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026929.1 | 1 | 767 | |||
GenBank mRNA | AL713776.1 | - | 1 | 766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
