siRNA Details
Gene Symbol: DKFZP586I1420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002186.1 | 1 | 1929 | |||
GenBank mRNA | AL050378.1 | - | 1 | 1929 | ||
BC064599.1 | - | 1 | 1241 | |||
BC073840.1 | - | 1 | 2113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
