siRNA Details
Gene Symbol: C20orf166-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033263.1 | 5 | 1025 | |||
GenBank mRNA | AK054875.1 | - | 3 | 683 | ||
AK056546.1 | - | 5 | 990 | |||
BC093856.1 | - | 1 | 227 | 7939701 | ||
BC111938.1 | - | 1 | 227 | 8327406 |
