siRNA Details
Gene Symbol: SNX32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055496.1 | - | 7 | 923 | ||
AK296055.1 | - | 4 | 770 | |||
AK309789.1 | - | 5 | 1179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
