siRNA Details
Gene Symbol: RALGPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152663.4 | NP_689876.2 | 14, 15 | 1611 | ||
XM_006711410.3 | XP_006711473.1 | 14, 15 | 1524 | |||
XM_006711411.3 | XP_006711474.1 | 7, 8 | 741 | |||
XM_011509688.1 | XP_011507990.1 | 14, 15 | 1525 | |||
GenBank mRNA | AK098470.1 | - | 3, 4 | 289 | ||
BC047391.1 | - | 14, 15 | 1596 | 5298449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
