siRNA Details
Gene Symbol: TBCEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130047.1 | NP_001123519.1 | 5 | 757 | ||
NM_152715.3 | NP_689928.3 | 5 | 884 | |||
XM_017017326.1 | XP_016872815.1 | 6 | 983 | |||
GenBank mRNA | AK295673.1 | - | 5 | 875 | ||
AK313324.1 | - | 5 | 779 | |||
BC020501.1 | - | 5 | 793 | |||
BC120988.1 | - | 4 | 707 | 40116354 | ||
BC120989.1 | - | 4 | 707 | 40116355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
