siRNA Details
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104452.1 | 25 | 2984 | |||
XM_005266861.2 | XP_005266918.1 | 23 | 2709 | |||
XM_011535569.1 | XP_011533871.1 | 23 | 2709 | |||
XM_011535570.1 | XP_011533872.1 | 24 | 2757 | |||
XM_011535571.2 | XP_011533873.1 | 24 | 3410 | |||
XM_011535574.1 | XP_011533876.1 | 23 | 2705 | |||
XM_011535575.2 | XP_011533877.1 | 24 | 2757 | |||
XM_011535576.2 | XP_011533878.1 | 24 | 2757 | |||
XM_011535580.2 | XP_011533882.1 | 14 | 1560 | |||
XM_011535582.2 | XP_011533884.1 | 25 | 2788 | |||
XM_017010397.1 | XP_016865886.1 | 24 | 2757 | |||
XM_017010398.1 | XP_016865887.1 | 24 | 3405 | |||
XM_017010400.1 | XP_016865889.1 | 23 | 2631 | |||
XM_017010402.1 | XP_016865891.1 | 23 | 2709 | |||
XM_017010403.1 | XP_016865892.1 | 24 | 2757 | |||
XM_017010404.1 | XP_016865893.1 | 9 | 980 | |||
GenBank mRNA | AK055461.1 | - | 6 | 531 | ||
AK125385.1 | - | 12 | 1419 | |||
AK131446.1 | - | 25 | 2964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
