siRNA Details
Gene Symbol: LINC00982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015440.1 | 3 | 3273 | |||
GenBank mRNA | AK124865.1 | - | 3 | 1789 | ||
AL713743.1 | - | 1 | 1270 | |||
AL833006.1 | - | 1 | 770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
