siRNA Details
Gene Symbol: FLJ23867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026900.1 | 1 | 1607 | |||
GenBank mRNA | AK074447.1 | - | 1 | 1627 | ||
AL832927.1 | - | 1 | 1607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
