siRNA Details
Gene Symbol: SAMD9L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303496.1 | NP_001290425.1 | 5 | 4942 | ||
NM_001303497.1 | NP_001290426.1 | 6 | 4771 | |||
NM_001303498.1 | NP_001290427.1 | 5 | 4507 | |||
NM_001303500.1 | NP_001290429.1 | 4 | 4375 | |||
NM_152703.3 | NP_689916.2 | 5 | 5147 | |||
XM_005250193.3 | XP_005250250.1 | 6 | 4579 | |||
XM_006715890.1 | XP_006715953.1 | 2 | 3948 | |||
XM_011515903.2 | XP_011514205.1 | 6 | 4832 | |||
XM_017011822.1 | XP_016867311.1 | 4 | 4896 | |||
XM_017011823.1 | XP_016867312.1 | 5 | 4652 | |||
GenBank mRNA | AB095926.1 | - | 5 | 5115 | ||
AF474973.1 | - | 6 | 4524 | |||
AK097204.1 | - | 1 | 1328 | |||
AK304307.1 | - | 1 | 3457 | |||
AK304779.1 | - | 1 | 2916 | |||
AL832264.1 | - | 1 | 2013 | |||
AL834232.1 | - | 1 | 655 | |||
BC029108.1 | - | 1 | 1433 | |||
BC038974.1 | - | 1 | 1351 | |||
BC127117.1 | - | 5 | 4384 | 40125349 | ||
BC127118.1 | - | 4 | 4759 | 40125356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
