siRNA Details
Gene Symbol: LOC100507431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125383.1 | 4 | 535 | |||
GenBank mRNA | BC031979.2 | - | 4 | 535 | ||
BP353837.1 | - | 2 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
