siRNA Details
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152730.5 | NP_689943.4 | 29 | 3356 | ||
NR_104452.1 | 33 | 4323 | ||||
XM_005266861.2 | XP_005266918.1 | 30 | 3479 | |||
XM_011535569.1 | XP_011533871.1 | 31 | 3542 | |||
XM_011535570.1 | XP_011533872.1 | 32 | 3590 | |||
XM_011535571.2 | XP_011533873.1 | 32 | 4243 | |||
XM_011535574.1 | XP_011533876.1 | 31 | 3538 | |||
XM_011535580.2 | XP_011533882.1 | 22 | 2393 | |||
XM_017010397.1 | XP_016865886.1 | 31 | 3527 | |||
XM_017010398.1 | XP_016865887.1 | 31 | 4175 | |||
XM_017010399.1 | XP_016865888.1 | 31 | 3467 | |||
XM_017010400.1 | XP_016865889.1 | 31 | 3464 | |||
XM_017010401.1 | XP_016865890.1 | 30 | 3404 | |||
XM_017010404.1 | XP_016865893.1 | 17 | 1813 | |||
GenBank mRNA | AK055461.1 | - | 13 | 1301 | ||
AK125385.1 | - | 19 | 2189 | |||
AK126259.1 | - | 9 | 925 | |||
AK131446.1 | - | 33 | 4303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
