siRNA Details
Gene Symbol: C6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK058086.1 | - | 7 | 932 | ||
FR832871.1 | - | 6 | 812 | |||
FR832872.1 | - | 5 | 797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
