siRNA Details
Gene Symbol: PAOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152911.3 | NP_690875.1 | 2, 3 | 757 | ||
NM_207127.2 | NP_997010.1 | 2, 3 | 757 | |||
NM_207128.2 | NP_997011.1 | 2, 3 | 757 | |||
NR_109763.1 | 2, 3 | 757 | ||||
GenBank mRNA | AF226657.1 | - | 1, 2 | 497 | ||
AF312698.1 | - | 2, 3 | 803 | |||
AK128001.1 | - | 2, 3 | 641 | |||
AY358418.1 | - | 2, 3 | 697 | |||
AY541513.1 | - | 2, 3 | 707 | |||
AY541515.1 | - | 2, 3 | 707 | |||
AY541516.1 | - | 2, 3 | 733 | |||
AY541519.1 | - | 2, 3 | 707 | |||
AY541520.1 | - | 1121 | ||||
AY541524.1 | - | 3, 4 | 607 | |||
BC032778.1 | - | 2, 3 | 768 | 4932036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
