siRNA Details
Gene Symbol: SRRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110199.2 | NP_001103669.1 | 15 | 2285 | ||
NM_001291831.1 | NP_001278760.1 | 16 | 2364 | |||
GenBank mRNA | AK092590.1 | - | 3 | 703 | ||
AK094397.1 | - | 1 | 896 | |||
BC063788.1 | - | 1 | 624 | |||
BC096755.1 | - | 4 | 814 |
