siRNA Details
Gene Symbol: BMP8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001720.3 | NP_001711.2 | 2 | 762 | ||
XM_005271149.4 | XP_005271206.1 | 2 | 480 | |||
XM_011542022.2 | XP_011540324.1 | 2 | 480 | |||
XM_011542024.2 | XP_011540326.2 | 2 | 480 | |||
XM_011542025.2 | XP_011540327.1 | 2 | 480 | |||
XM_017002155.1 | XP_016857644.1 | 2 | 480 | |||
XM_017002156.1 | XP_016857645.1 | 2 | 480 | |||
GenBank mRNA | AY303955.1 | - | 2 | 762 | ||
BC023526.2 | - | 2 | 361 | |||
BC108678.1 | - | 2 | 750 | 6045180 |
Gene Symbol: BMP8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181809.3 | NP_861525.2 | 2 | 732 | ||
XM_006710616.3 | XP_006710679.1 | 2 | 752 | |||
XM_011541381.2 | XP_011539683.1 | 2 | 749 | |||
XM_011541382.2 | XP_011539684.1 | 2 | 749 | |||
XM_017001198.1 | XP_016856687.1 | 2 | 749 | |||
GenBank mRNA | AY303954.1 | - | 2 | 732 | ||
BC131617.1 | - | 2 | 48 | 40116584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
