siRNA Details
Gene Symbol: NAGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153006.2 | NP_694551.1 | 7 | 1565 | ||
XM_011524438.1 | XP_011522740.1 | 6 | 1500 | |||
XM_011524439.2 | XP_011522741.1 | 7 | 1455 | |||
GenBank mRNA | AK074407.1 | - | 7 | 1495 | ||
AK091938.1 | - | 4 | 1453 | |||
AK300923.1 | - | 8 | 1471 | |||
AK314432.1 | - | 7 | 1683 | |||
AY116538.1 | - | 7 | 1565 | |||
AY158070.1 | - | 7 | 1286 | |||
BC111713.1 | - | 8 | 1351 | 40005903 |
