siRNA Details
Gene Symbol: ST6GALNAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160011.1 | NP_001153483.1 | 3 | 606 | ||
NM_152996.2 | NP_694541.2 | 3 | 606 | |||
XM_006710553.3 | XP_006710616.1 | 4 | 602 | |||
XM_017000937.1 | XP_016856426.1 | 3 | 3748 | |||
XM_017000938.1 | XP_016856427.1 | 3 | 3760 | |||
XM_017000939.1 | XP_016856428.1 | 3 | 3758 | |||
XM_017000940.1 | XP_016856429.1 | 4 | 865 | |||
XM_017000941.1 | XP_016856430.1 | 3 | 1520 | |||
XM_017000942.1 | XP_016856431.1 | 3 | 1325 | |||
GenBank mRNA | AJ507291.1 | - | 3 | 464 | ||
AK091215.1 | - | 3 | 591 | |||
AK297363.1 | - | 2 | 375 | |||
AY358540.1 | - | 3 | 565 | |||
BC039520.1 | - | 2 | 317 | |||
BC059363.1 | - | 3 | 577 | 30341233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
