siRNA Details
Gene Symbol: OFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF520800.1 | - | 7, 8 | 673 | ||
AF520805.1 | - | 5, 6 | 548 | |||
AF548115.1 | - | 2, 3 | 244 | |||
AF548116.1 | - | 3, 4 | 330 | |||
AF548121.1 | - | 240 | ||||
BC113541.1 | - | 6, 7 | 612 | 8322593 | ||
BC113543.1 | - | 6, 7 | 612 | 8322595 | ||
BC143837.1 | - | 7, 8 | 624 | 9052352 |
Gene Symbol: LOC107983965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011612.1 | XP_016867101.1 | 8, 9 | 970 |
