siRNA Details
Gene Symbol: LOC149373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256615.1 | NP_001243544.1 | 3 | 636 | ||
GenBank mRNA | AK054992.1 | - | 1 | 1588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
