siRNA Details
Gene Symbol: PLXNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025179.3 | NP_079455.3 | 32 | 10605 | ||
XM_005273164.2 | XP_005273221.1 | 33 | 10864 | |||
GenBank mRNA | AK055196.1 | - | 1 | 1983 | ||
AK094259.1 | - | 1 | 807 | |||
AK125154.1 | - | 1 | 2533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
