siRNA Details
Gene Symbol: LINC00889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026935.1 | 1 | 1260 | |||
GenBank mRNA | AK055234.1 | - | 1 | 809 | ||
AK055382.1 | - | 1 | 944 | |||
BC105719.1 | - | 1 | 934 | |||
BC105725.1 | - | 1 | 934 | |||
BC105726.1 | - | 1 | 934 | |||
BC105727.1 | - | 1 | 934 | |||
BC139831.1 | - | 1 | 937 |
Gene Symbol: GADL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832766.1 | - | 12 | 3257 |
Gene Symbol: LINC00652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094606.1 | - | 3 | 2651 |
Gene Symbol: TSKU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318477.1 | NP_001305406.1 | 1 | 419 | ||
GenBank mRNA | AK126536.1 | - | 1 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
