siRNA Details
Gene Symbol: SPATA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166271.2 | NP_001159743.1 | 13 | 5206 | ||
NM_001286792.1 | NP_001273721.1 | 15 | 5530 | |||
NM_001286793.1 | NP_001273722.1 | 9 | 2754 | |||
NM_001286794.1 | NP_001273723.1 | 11 | 3148 | |||
NM_001286795.1 | NP_001273724.1 | 10 | 2940 | |||
NM_153023.3 | NP_694568.1 | 12 | 3442 | |||
GenBank mRNA | AK055770.1 | - | 12 | 3410 | ||
AK074117.1 | - | 3 | 1652 | |||
AK097217.1 | - | 6 | 2083 | |||
AK123031.1 | - | 10 | 2840 | |||
AK160371.1 | - | 9 | 2629 | |||
BX648244.1 | - | 15 | 5352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
