siRNA Details
Gene Symbol: ZNF524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153219.3 | NP_694951.1 | 2 | 942 | ||
XM_011526487.2 | XP_011524789.1 | 2 | 1183 | |||
GenBank mRNA | BC007396.2 | - | 1 | 825 | ||
BC014666.1 | - | 1 | 885 | 4864278 | ||
BC067748.1 | - | 2 | 913 | 5313025 |
