siRNA Details
Gene Symbol: LOC439933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122080.1 | 1 | 132 | |||
GenBank mRNA | BC030111.1 | - | 1 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
