siRNA Details
Gene Symbol: ZDHHC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286638.1 | NP_001273567.1 | 11 | 2839 | ||
NM_153251.3 | NP_694983.2 | 12 | 2862 | |||
NR_104486.1 | 15 | 3107 | ||||
NR_104487.1 | 13 | 2911 | ||||
XM_006719795.1 | XP_006719858.1 | 13 | 2883 | |||
XM_006719796.1 | XP_006719859.1 | 13 | 2880 | |||
XM_006719797.1 | XP_006719860.1 | 12 | 2805 | |||
XM_011535030.2 | XP_011533332.1 | 7 | 2362 | |||
XM_017020525.1 | XP_016876014.1 | 11 | 2576 | |||
XM_017020526.1 | XP_016876015.1 | 14 | 2649 | |||
XM_017020527.1 | XP_016876016.1 | 12 | 2781 | |||
GenBank mRNA | AK098818.1 | - | 1 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
